2,2'-Furil dioxime 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR400953-5G
OR400953-1G
APOSOR400953-5GEA
543
EUR
APOSOR400953-5G
APOSOR400953-1G
2,2'-Furil dioxime 97%
2,2'-Furil dioxime
Mixture of isomers
Formel:
C₁₀H₈N₂O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00075315 CAS-Nummer: 522-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...