1-Brom-2-chlor-4-iodbenzol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR400630-100G
OR400630-25G
OR400630-5G
APOSOR400630-100GEA
112
EUR
APOSOR400630-100G
APOSOR400630-25G
APOSOR400630-5G
1-Brom-2-chlor-4-iodbenzol 98%
1-Bromo-2-chloro-4-iodobenzene
Formel:
C₆H₃BrCl Schmelzpunkt: 38…39 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04973904 CAS-Nummer: 535934-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...