7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-[1,2,4]triazolo[1,5-a]pyridin-2-amine

OR400610-1G OR400610-5G
APOSOR400610-1GEA 108 EUR
APOSOR400610-1G APOSOR400610-5G
7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
7-Bromo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Formel: C₆H₅BrN₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD11846606
CAS-Nummer: 882521-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR