3-(Dimethylamino)benzoesäure
Lieferant: Apollo Scientific
OR400573-100G
OR400573-500G
APOSOR400573-100GEA
24.4
EUR
APOSOR400573-100G
APOSOR400573-500G
3-(Dimethylamino)benzoesäure
3-(Dimethylamino)benzoesäure
Formel:
C₉H₁₁NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00002497 CAS-Nummer: 99-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...