5,7-Dichlor-4-chinolinol 97%
Lieferant: Apollo Scientific
Synonyme:
5,7-Dichloro-4-hydroxyquinoline, 5,7-Dichloroquinolin-4-ol
OR3930-1G
OR3930-500MG
APOSOR3930-1GEA
139
EUR
APOSOR3930-1G
APOSOR3930-500MG
5,7-Dichlor-4-chinolinol 97%
5,7-Dichlor-4-chinolinol
Formel:
C₉H₅Cl₂NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00219834 CAS-Nummer: 171850-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...