2-(Morpholino)phenylboronsäure 98%
Lieferant: Apollo Scientific
Synonyme:
(2-Morpholinophenyl)boronic acid
OR360301-250MG
OR360301-1G
OR360301-5G
APOSOR360301-250MGEA
94.4
EUR
APOSOR360301-250MG
APOSOR360301-1G
APOSOR360301-5G
2-(Morpholino)phenylboronsäure 98%
2-(Morpholino)phenylboronsäure
Formel:
C₁₀H₁₄BNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03095109 CAS-Nummer: 933052-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...