2-Isopropoxy-3-pyridinboronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Isopropoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine, 2-Isopropoxypyridine-3-boronic acid pinacol ester, 2-Isopropoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR360051-1G
OR360051-5G
OR360051-25G
APOSOR360051-1GEA
95.7
EUR
APOSOR360051-1G
APOSOR360051-5G
APOSOR360051-25G
2-Isopropoxy-3-pyridinboronsäurepinakolester
2-Isopropoxy-3-pyridinboronsäurepinakolester
Formel:
C₁₄H₂₂BNO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD07366722 CAS-Nummer: 848243-25-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...