5-Acetyl-2-norbornene
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-Norbornen-2-yl Ketone
OR350014-1G
OR350014-5G
APOSOR350014-1GEA
43.5
EUR
APOSOR350014-1G
APOSOR350014-5G
5-Acetyl-2-norbornene
5-Acetyl-2-norbornene
Formel:
C₉H₁₂O Molekulargewicht: 136.19 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00167570 CAS-Nummer: 5063-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...