2-Amino-6-brom-5-methoxypyridin

Lieferant: Apollo Scientific

Synonyme: 6-Bromo-5-methoxy-pyridin-2-ylamine, 6-Bromo-5-methoxypyridin-2-amine

OR346242-1G OR346242-500MG
APOSOR346242-1GEA 369 EUR
APOSOR346242-1G APOSOR346242-500MG
2-Amino-6-brom-5-methoxypyridin
2-Amino-6-bromo-5-methoxypyridine
Formel: C₆H₇BrN₂O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD06662268
CAS-Nummer: 79491-43-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR