Ethyl-5-hydroxymethyl-3-methylisoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR346090-1G OR346090-5G
APOSOR346090-1GEA 0 EUR
APOSOR346090-1G APOSOR346090-5G
Ethyl-5-hydroxymethyl-3-methylisoxazole-4-carboxylate
Ethyl-5-hydroxymethyl-3-methylisoxazole-4-carboxylate
Formel: C₈H₁₁NO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10568348
CAS-Nummer: 95104-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR