Ethyl-3-chloro-2,4-dioxopentanoate

Lieferant: Apollo Scientific

OR346085-1G OR346085-5G OR346085-10G
APOSOR346085-1GEA 0 EUR
APOSOR346085-1G APOSOR346085-5G APOSOR346085-10G
Ethyl-3-chloro-2,4-dioxopentanoate
Ethyl-3-chloro-2,4-dioxopentanoate
Formel: C₇H₉ClO₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00173820
CAS-Nummer: 34959-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR