Ethyl-2-brom-5-oxazolcarboxylat
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-bromooxazole-5-carboxylate
OR345708-1G
OR345708-250MG
OR345708-5G
APOSOR345708-1GEA
151
EUR
APOSOR345708-1G
APOSOR345708-250MG
APOSOR345708-5G
Ethyl-2-brom-5-oxazolcarboxylat
Ethyl-2-brom-5-oxazolcarboxylat
Formel:
C₆H₆BrNO₃ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09743496 CAS-Nummer: 1060816-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...