2-Bromo-3-nitropyridine-4-amine 97%
Lieferant: Apollo Scientific
Synonyme:
4-Amino-2-bromo-3-nitropyridine
OR345421-1G
OR345421-100MG
OR345421-250MG
APOSOR345421-1GEA
215
EUR
APOSOR345421-1G
APOSOR345421-100MG
APOSOR345421-250MG
2-Bromo-3-nitropyridine-4-amine 97%
2-Bromo-3-nitropyridine-4-amine
Formel:
C₅H₄BrN₃O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD11656226 CAS-Nummer: 84487-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...