2,5-Dichlor-4-iodpyridin 98%
Lieferant: Apollo Scientific
OR345414-10G
OR345414-25G
OR345414-5G
APOSOR345414-10GEA
177
EUR
APOSOR345414-10G
APOSOR345414-25G
APOSOR345414-5G
2,5-Dichlor-4-iodpyridin 98%
2,5-Dichloro-4-iodopyridine
Formel:
C₅H₂Cl₂IN Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD13185538 CAS-Nummer: 796851-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...