4-Amino-3-brom-5-iodpyridin 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-iodopyridin-4-amine, 3-Bromo-5-iodo-4-pyridylamine

OR345153-1G OR345153-10G OR345153-5G
APOSOR345153-1GEA 94.4 EUR
APOSOR345153-1G APOSOR345153-10G APOSOR345153-5G
4-Amino-3-brom-5-iodpyridin 95+%
4-Amino-3-bromo-5-iodopyridine
Formel: C₅H₄BrIN₂
Molekulargewicht: 298.91 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07781147
CAS-Nummer: 902837-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR