Sulphapyridine 98+%

Lieferant: Apollo Scientific

OR345024-100G OR345024-25G
APOSOR345024-100GEA 43.5 EUR
APOSOR345024-100G APOSOR345024-25G
Sulphapyridine 98+%
Sulphapyridine
Formel: C₁₁H₁₁N₃O₂S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00038036
CAS-Nummer: 144-83-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR