(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol 95%
Lieferant: Apollo Scientific
OR33483-1G
OR33483-5G
APOSOR33483-1GEA
55.1
EUR
APOSOR33483-1G
APOSOR33483-5G
(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol 95%
(2-(1H-1,2,4-Triazol-1-yl)phenyl)methanol
Formel:
C₉H₉N₃O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05864820 CAS-Nummer: 914349-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...