2-(4-Formylphenoxy)acetonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR33278-1G
OR33278-25G
OR33278-5G
APOSOR33278-1GEA
180
EUR
APOSOR33278-1G
APOSOR33278-25G
APOSOR33278-5G
2-(4-Formylphenoxy)acetonitrile 95%
2-(4-Formylphenoxy)acetonitrile
Formel:
C₉H₇NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02629373 CAS-Nummer: 385383-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...