7-Cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile 95%

Lieferant: Apollo Scientific

OR33159-1G OR33159-5G
APOSOR33159-1GEA 270 EUR
APOSOR33159-1G APOSOR33159-5G
7-Cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile 95%
7-Cyclopropylpyrazolo[1,5-a]pyrimidine-3-carbonitrile
Formel: C₁₀H₈N₄
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03305760
CAS-Nummer: 866132-04-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR