N-(4-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide 95%

Lieferant: Apollo Scientific

OR33106-1G OR33106-25G OR33106-5G
APOSOR33106-1GEA 180 EUR
APOSOR33106-1G APOSOR33106-25G APOSOR33106-5G
N-(4-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide 95%
N-(4-Acetylphenyl)-3-chloro-2,2-dimethylpropanamide
Formel: C₁₃H₁₆ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00793413
CAS-Nummer: 339015-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR