4-[(3,5-Dimethyl-4-isoxazolyl)methyl]benzenol 95%
Lieferant: Apollo Scientific
OR33060-1G
OR33060-25G
OR33060-5G
APOSOR33060-1GEA
122
EUR
APOSOR33060-1G
APOSOR33060-25G
APOSOR33060-5G
4-[(3,5-Dimethyl-4-isoxazolyl)methyl]benzenol 95%
4-[(3,5-Dimethyl-4-isoxazolyl)methyl]benzenol
Formel:
C₁₂H₁₃NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00215380 CAS-Nummer: 75999-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...