4-(tert-Butyl)cyclohexanonoxim 95%

Lieferant: Apollo Scientific

Synonyme: N-(4-tert-Butylcyclohexylidene)hydroxylamine

OR32681-1G OR32681-25G OR32681-5G
APOSOR32681-1GEA 180 EUR
APOSOR32681-1G APOSOR32681-25G APOSOR32681-5G
4-(tert-Butyl)cyclohexanonoxim 95%
4-(tert-Butyl)cyclohexanonoxim
Formel: C₁₀H₁₉NO
Lagerungstemperatur: Raumtemperatur
CAS-Nummer: 4701-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR