Methyl-6-(1H-imidazol-1-yl)nicotinate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-(1H-imidazol-1-yl)pyridine-3-carboxylate
OR32528-1G
OR32528-5G
APOSOR32528-1GEA
180
EUR
APOSOR32528-1G
APOSOR32528-5G
Methyl-6-(1H-imidazol-1-yl)nicotinate 95%
Methyl-6-(1H-imidazol-1-yl)nicotinate
Formel:
C₁₀H₉N₃O₂ Molekulargewicht: 203.2 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00140820 CAS-Nummer: 111205-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...