N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Succinimidyl (Boc-aminooxy)acetate, 2,5-Dioxopyrrolidin-1-yl 2-(((tert-butoxycarbonyl)amino)oxy)acetate, (Boc-aminooxy)acetic acid N-Succinimidyl ester, [(tert-Butoxycarbonyl)aminooxy]acetic acid N-Succinimidyl ester, Boc-NH-O-C1-NHS ester
OR323004-1G
OR323004-5G
APOSOR323004-1GEA
157
EUR
APOSOR323004-1G
APOSOR323004-5G
N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate 98%
N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate
Formel:
C₁₁H₁₆N₂O₇ Molekulargewicht: 288.26 g/mol Schmelzpunkt: 108…110 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD18382162 CAS-Nummer: 80366-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...