1,2,4-Triaminobenzene dihydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2,4-Benzenetriamine Dihydrochloride
OR322745-25G
OR322745-5G
APOSOR322745-25GEA
197
EUR
APOSOR322745-25G
APOSOR322745-5G
1,2,4-Triaminobenzene dihydrochloride
1,2,4-Triaminobenzene dihydrochloride
Formel:
C₆H₉N₃ Schmelzpunkt: 290 °C (dec.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00016619 CAS-Nummer: 615-47-4 EINECS: 210-428-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...