6-(1H-Pyrrol-1-yl)nicotinonitrile 95%
Lieferant: Apollo Scientific
Synonyme:
6-(1H-Pyrrol-1-yl)pyridine-3-carbonitrile
OR32272-1G
OR32272-5G
OR32272-25G
APOSOR32272-1GEA
180
EUR
APOSOR32272-1G
APOSOR32272-5G
APOSOR32272-25G
6-(1H-Pyrrol-1-yl)nicotinonitrile 95%
6-(1H-Pyrrol-1-yl)nicotinonitrile
Formel:
C₁₀H₇N₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00828638 CAS-Nummer: 218157-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...