3-(Dimethylamino)propanoic acid hydrochloride
Lieferant: Apollo Scientific
Synonyme:
3-(Dimethylamino)propionic Acid HCl, 3-(Dimethylamino)propionic acid hydrochloride, N,N-Dimethyl-beta-alanine hydrochloride
OR322402-25G
OR322402-100G
APOSOR322402-25GEA
69.6
EUR
APOSOR322402-25G
APOSOR322402-100G
3-(Dimethylamino)propanoic acid hydrochloride
3-(Dimethylamino)propanoic acid hydrochloride
Formel:
C5H12ClNO2 Schmelzpunkt: 189 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02093464 CAS-Nummer: 14788-12-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...