1-(3-Piperidinyl)methanamin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Piperidin-3-ylmethanamine

OR321303-1G OR321303-250MG
APOSOR321303-1GEA 53.8 EUR
APOSOR321303-1G APOSOR321303-250MG
1-(3-Piperidinyl)methanamin 97%
3-(Aminomethyl)piperidine
Formel: C₆H₁₄N₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD03427344
CAS-Nummer: 23099-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR