(1-Methyl-1H-pyrazol-4-yl)methylamine

Lieferant: Apollo Scientific

Synonyme: (1-Methyl-1H-pyrazol-4-yl)methanamine

OR318083-10G OR318083-1G OR318083-5G
APOSOR318083-10GEA 444 EUR
APOSOR318083-10G APOSOR318083-1G APOSOR318083-5G
(1-Methyl-1H-pyrazol-4-yl)methylamine
(1-Methyl-1H-pyrazol-4-yl)methylamine
Formel: C₅H₉N₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02090924
CAS-Nummer: 400877-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR