(S)-1-Boc-2-azetidinemethanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-1-Boc-2-Azetidinemethanol, (S)-1-Boc-2-azetidinemethanol, (S)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol
OR317181-1G
OR317181-5G
APOSOR317181-1GEA
119
EUR
APOSOR317181-1G
APOSOR317181-5G
(S)-1-Boc-2-azetidinemethanol
(S)-1-Boc-2-azetidinemethanol
Formel:
C₉H₁₇NO₃ Dichte: 1.05 g/cm³ (25 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03093620 CAS-Nummer: 161511-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...