3-(Diethylboryl)pyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Diethylboryl)pyridine
OR315000-100G
OR315000-25G
APOSOR315000-100GEA
209
EUR
APOSOR315000-100G
APOSOR315000-25G
3-(Diethylboryl)pyridin
Diethyl(3-pyridyl)borane
Formel:
C₉H₁₄BN Schmelzpunkt: 172…175 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012348 CAS-Nummer: 89878-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...