(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3R,4R)-tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate, trans-3-Amino-4-hydroxypyrrolidine, N1-BOC protected
OR313021-1G
OR313021-250MG
APOSOR313021-1GEA
78.3
EUR
APOSOR313021-1G
APOSOR313021-250MG
(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate
(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate
Formel:
C₉H₁₈N₂O₃ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD11111251 CAS-Nummer: 330681-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...