4-N-Boc-amino-4-carboxytetrahydropyran 97%

Lieferant: Apollo Scientific

Synonyme: 4-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid

OR312142-1G OR312142-5G
APOSOR312142-1GEA 75.4 EUR
APOSOR312142-1G APOSOR312142-5G
4-N-Boc-amino-4-carboxytetrahydropyran 97%
4-N-Boc-amino-4-carboxytetrahydropyran
Formel: C₁₁H₁₉NO₅
Molekulargewicht: 245.275 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD02683136
CAS-Nummer: 172843-97-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR