3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR312112-1G OR312112-5G
APOSOR312112-1GEA 82.8 EUR
APOSOR312112-1G APOSOR312112-5G
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
3-(Methoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₈H₁₀O₄
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD20638314
CAS-Nummer: 83249-10-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR