3'-Chlor-2-chloracetanilid

Lieferant: Apollo Scientific
Achtung

OR3114-1G OR3114-5G
APOSOR3114-1GEA 36.4 EUR
APOSOR3114-1G APOSOR3114-5G
3'-Chlor-2-chloracetanilid
3'-Chlor-2-chloracetanilid
Formel: C₈H₇Cl₂NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00018896
CAS-Nummer: 2564-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR