4-Hydrazinopiperidine dihydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-Hydrazinylpiperidine dihydrochloride

OR310384-1G OR310384-500MG
APOSOR310384-1GEA 894 EUR
APOSOR310384-1G APOSOR310384-500MG
4-Hydrazinopiperidine dihydrochloride
4-Hydrazinopiperidine dihydrochloride
Formel: C₅H₁₅Cl₂N₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD18071345
CAS-Nummer: 380226-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR