4-Amino-3-bromchinolin

Lieferant: Apollo Scientific

OR309338-1G OR309338-5G
APOSOR309338-1GEA 484 EUR
APOSOR309338-1G APOSOR309338-5G
4-Amino-3-bromchinolin
4-Amino-3-bromoquinoline
Formel: C₉H₇BrN₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00234502
CAS-Nummer: 36825-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR