7-Boc-2-oxa-7-azaspiro[3.5]nonane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-oxa-7-azaspiro[3.5]nonane-7-carboxylate, 2-Oxa-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

OR309124-1G OR309124-250MG OR309124-5G
APOSOR309124-1GEA 77 EUR
APOSOR309124-1G APOSOR309124-250MG APOSOR309124-5G
7-Boc-2-oxa-7-azaspiro[3.5]nonane 97%
7-Boc-2-oxa-7-azaspiro[3.5]nonane
Formel: C₁₂H₂₁NO₃
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD11035887
CAS-Nummer: 240401-27-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR