Azetidin-3-amine dihydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Aminoazetidine dihydrochloride

OR309033-1G OR309033-5G OR309033-25G
APOSOR309033-1GEA 84.1 EUR
APOSOR309033-1G APOSOR309033-5G APOSOR309033-25G
Azetidin-3-amine dihydrochloride 97%
Azetidin-3-amine dihydrochloride
Formel: C₃H₁₀Cl₂N₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09910173
CAS-Nummer: 102065-89-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR