(R)-3-Methylmorpholine
Lieferant: Apollo Scientific
|
Achtung
|
OR308175-1G
OR308175-5G
OR308175-10G
APOSOR308175-1GEA
21.9
EUR
APOSOR308175-1G
APOSOR308175-5G
APOSOR308175-10G
(R)-3-Methylmorpholine
(R)-3-Methylmorpholine
Formel:
C₅H₁₁NO Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD10698482 CAS-Nummer: 74572-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...