Cyclobutylmethanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR308087-1G OR308087-5G
APOSOR308087-1GEA 21.9 EUR
APOSOR308087-1G APOSOR308087-5G
Cyclobutylmethanamine hydrochloride
Cyclobutylmethanamine hydrochloride
Formel: C₅H₁₂ClN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00034952
CAS-Nummer: 5454-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR