3-Hydroxy-1-methylazetidine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methylazetidin-3-ol, 1-Methyl-3-azetidinol

OR308027-250MG OR308027-1G
APOSOR308027-250MGEA 148 EUR
APOSOR308027-250MG APOSOR308027-1G
3-Hydroxy-1-methylazetidine 96%
3-Hydroxy-1-methylazetidine
Formel: C₄H₉NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD10565802
CAS-Nummer: 111043-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR