1-Boc-3-(iodomethyl)azetidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Boc-3-(iodomethyl)azetidine, tert-Butyl 3-(iodomethyl)azetidine-1-carboxylate
OR308023-1G
OR308023-5G
APOSOR308023-1GEA
24.8
EUR
APOSOR308023-1G
APOSOR308023-5G
1-Boc-3-(iodomethyl)azetidine
1-Boc-3-(iodomethyl)azetidine
Formel:
C₉H₁₆INO₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD09951816 CAS-Nummer: 253176-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...