tert-Butyl-3-amino-3-methylazetidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-amino-3-methylazetidine-1-carboxylate

OR308015-1G OR308015-5G
APOSOR308015-1GEA 130 EUR
APOSOR308015-1G APOSOR308015-5G
tert-Butyl-3-amino-3-methylazetidine-1-carboxylate
tert-Butyl-3-amino-3-methylazetidine-1-carboxylate
Formel: C₉H₁₈N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12408556
CAS-Nummer: 1158758-77-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR