(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR307061-5G
OR307061-100G
APOSOR307061-5GEA
26.1
EUR
APOSOR307061-5G
APOSOR307061-100G
(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid
(R)-3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid
Formel:
C₈H₁₆N₂O₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01632072 CAS-Nummer: 76387-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...