4-Brom-2-nitroanilin
Lieferant: Apollo Scientific
|
Achtung
|
OR3065-100G
OR3065-250G
OR3065-500G
APOSOR3065-100GEA
46.4
EUR
APOSOR3065-100G
APOSOR3065-250G
APOSOR3065-500G
4-Brom-2-nitroanilin
4-Brom-2-nitroanilin
Formel:
C₆H₅BrN₂O₂ Schmelzpunkt: 110…114 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00041312 CAS-Nummer: 875-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...