7-Bromimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR30603-1G
OR30603-250MG
APOSOR30603-1GEA
55.1
EUR
APOSOR30603-1G
APOSOR30603-250MG
7-Bromimidazo[1,2-a]pyridin
7-Bromimidazo[1,2-a]pyridin
Formel:
C₇H₅BrN₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07778353 CAS-Nummer: 808744-34-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...