6-Chloro-5-iodopyridine-3-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-5-iodonicotinonitrile
OR303862-1G
OR303862-500MG
APOSOR303862-1GEA
894
EUR
APOSOR303862-1G
APOSOR303862-500MG
6-Chloro-5-iodopyridine-3-carbonitrile
6-Chloro-5-iodopyridine-3-carbonitrile
Formel:
C₆H₂ClIN₂ Schmelzpunkt: 150…152 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD16710295 CAS-Nummer: 856213-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...