5-Brom-2-chlor-4-iodpyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR303701-1G
OR303701-5G
APOSOR303701-1GEA
21.5
EUR
APOSOR303701-1G
APOSOR303701-5G
5-Brom-2-chlor-4-iodpyridin
5-Brom-2-chlor-4-iodpyridin
Formel:
C₅H₂BrClIN Schmelzpunkt: 149…150 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09972189 CAS-Nummer: 401892-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...