3-[(7-Chloroquinolin-4-yl)amino]phenol hydrochloride

Lieferant: Apollo Scientific

Synonyme: 6-[(2R,6S)-2,6-Dimethylmorpholin-4-yl]pyridin-3-amine dihydrochloride

OR303606-1G OR303606-500MG OR303606-5G
APOSOR303606-1GEA 270 EUR
APOSOR303606-1G APOSOR303606-500MG APOSOR303606-5G
3-[(7-Chloroquinolin-4-yl)amino]phenol hydrochloride
3-[(7-Chloroquinolin-4-yl)amino]phenol hydrochloride
Formel: C₁₅H₁₂Cl₂N₂O
Schmelzpunkt: 237…239 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD14155833
CAS-Nummer: 154179-39-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR